Tin ≥99.8% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
43236.GH 43236.HP 43236.FI
43236.GHEA 213 EUR
43236.GH 43236.HP 43236.FI
Tin ≥99.8% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Tin
Purity: 99.8% CAS No.: 7440-31-5 Physical Form: ≈36g/50x50mm Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587