Bismuth ≥99.99% (metals basis), rods, Ø 11 mm (0.43 in)
Supplier: Thermo Fisher Scientific
41534.14 41534.22 41534.36
41534.14EA 96 EUR
41534.14 41534.22 41534.36
Bismuth ≥99.99% (metals basis), rods, Ø 11 mm (0.43 in)
Bismuth
Purity: 99.99% CAS No.: 7440-69-9 Physical Form: ≈9.3g/cm Odour: Odourless MDL No.: MFCD00134033
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297