Sulphur ≥99.9995% (metals basis), pieces, Puratronic®
Supplier: Thermo Fisher Scientific
Danger
10343.14 10343.22 10343.36
10343.14EA 104 EUR
10343.14 10343.22 10343.36
Sulphur ≥99.9995% (metals basis), pieces, Puratronic®
Sulphur
Purity: 99.9995% CAS No.: 7704-34-9 Physical Form: Random Sizes Odour: Weak UN No.: 1350 MDL No.: MFCD00085316
Formula: S
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 113…119 °C
Density: 2.36 g/cm³ (20 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059