Sulphur ≥99%, flower
Supplier: Thermo Fisher Scientific
Warning
A17260.22 A17260.0E A17260.36
A17260.22EA 25.4 EUR
A17260.22 A17260.0E A17260.36
Sulphur ≥99%, flower
Sulphur
Formula: S
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 113…119 °C
Density: 2.36 g/cm³ (20 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059