Iodine, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
229695-100G 229695-20G
SIAL229695-100GEA 482 EUR
SIAL229695-100G SIAL229695-20G
Iodine, Sigma-Aldrich®
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 4.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036