Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
40288.DH 40288.DJ 40288.HF
40288.DHEA 900 EUR
40288.DH 40288.DJ 40288.HF
Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Erbium
Purity: 99.9% CAS No.: 7440-52-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈1.2g/mm, Cast rod, diameter is approximate Odour: Odourless
Formula: Er
MW: 167.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1