Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Danger
Packaged under argon.
12395.FF 12395.FI
12395.FFEA 179 EUR
12395.FF 12395.FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Gadolinium
Purity: 99.9% CAS No.: 7440-54-2 Physical Form: ≈1.2g/25x25mm, Packaged under argon UN No.: 3178 MDL No.: MFCD00011022
Formula: Gd
MW: 157.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2
UN: 3178
ADR: 4.1 II