Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
40289.DH 40289.DJ 40289.HF
40289.DHEA 522 EUR
40289.DH 40289.DJ 40289.HF
Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Gadolinium
Purity: 99.9% CAS No.: 7440-54-2 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.25g/mm, Cast rod, diameter is approximate MDL No.: MFCD00011022
Formula: Gd
MW: 157.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2