Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
40290.DH 40290.HF
40290.DHEA 713 EUR
40290.DH 40290.HF
Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Gadolinium
Purity: 99.9% CAS No.: 7440-54-2 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.97g/mm, Cast rod, diameter is approximate MDL No.: MFCD00011022
Formula: Gd
MW: 157.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2