Gold 99,9985 Metals basis, wire, Premion®, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
10966.BQ 10966.BY 10966.CB 10966.BU
10966.BQEA 63.1 EUR
10966.BQ 10966.BY 10966.CB 10966.BU
Gold 99,9985 Metals basis, wire, Premion®, Ø 0.5 mm (0.02 in)
Gold
Purity: 99.9985% CAS No.: 7440-57-5 Physical Form: ≈3.92g/m Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526