Gold ≥99.998% (metals basis), wire, Premion®, Ø 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
10968.G1 10968.G5 10968.G9
10968.G1EA 101 EUR
10968.G1 10968.G5 10968.G9
Gold ≥99.998% (metals basis), wire, Premion®, Ø 0.1 mm (0.004 in)
Gold
Purity: 99.998% CAS No.: 7440-57-5 Physical Form: ≈0.15g/m Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526