Gold ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
14721.FF 14721.FI 14721.GH
14721.FFEA 406 EUR
14721.FF 14721.FI 14721.GH
Gold ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Gold
Purity: 99.95% CAS No.: 7440-57-5 Physical Form: ≈1.2g/25x25mm Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526