Gold 99,99 Metals basis, gauze_wire 52 mesh, Ø 0.102 mm (0.004 in)
Supplier: Thermo Fisher Scientific
40930.GH 40930.FF 40930.FI 40930.FY
40930.GHEA 2980 EUR
40930.GH 40930.FF 40930.FI 40930.FY
Gold 99,99 Metals basis, gauze_wire 52 mesh, Ø 0.102 mm (0.004 in)
Gold
Purity: 99.99% CAS No.: 7440-57-5 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00003436
Electrodes
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526