Gold, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
265780-2.8G
265780-280MG
SIAL265780-2.8GEA
781
EUR
SIAL265780-2.8G
SIAL265780-280MG
Gold, Sigma-Aldrich®
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...