Gold, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
265810-1.2G 265810-4.8G
SIAL265810-1.2GEA 462 EUR
SIAL265810-1.2G SIAL265810-4.8G
Gold, Sigma-Aldrich®
Gold
Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526