Gold, powder, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

326585-500MG 326585-5G
SIAL326585-500MGEA 200 EUR
SIAL326585-500MG SIAL326585-5G
Gold, powder, Sigma-Aldrich®
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR