Copper, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
254177-125G
254177-25G
SIAL254177-125GEA
631
EUR
SIAL254177-125G
SIAL254177-25G
Copper, Sigma-Aldrich®
Copper
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |