Copper, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

326488-250G 326488-50G
SIAL326488-250GEA 708 EUR
SIAL326488-250G SIAL326488-50G
Copper, Sigma-Aldrich®
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR