Copper, powder, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
266086-25G
266086-2KG
266086-500G
SIAL266086-25GEA
38
EUR
SIAL266086-25G
SIAL266086-2KG
SIAL266086-500G
Copper, powder, Sigma-Aldrich®
Copper
Formula:
Cu Boiling Pt: 2567 °C (1013 hPa) Melting Pt: 1083.4 °C Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 UN: 3089 ADR: 4.1,III |