Copper, powder, Sigma-Aldrich®
Supplier: Merck
Warning
MQ100
326453-1KG 326453-250G
SIAL326453-1KGEA 259 EUR
SIAL326453-1KG SIAL326453-250G
Copper, powder, Sigma-Aldrich®
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545