Silver ≥99.95% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Fisher Scientific
12190.GT 12190.GV 12190.IA 12190.1X
12190.GTEA 118 EUR
12190.GT 12190.GV 12190.IA 12190.1X
Silver ≥99.95% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)
Silver
Purity: 99.95% CAS No.: 7440-22-4 Physical Form: ≈5.9g/150x150mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647