Silver ≥99.9% (metals basis), flakes 80% <20 µm
Supplier: Thermo Fisher Scientific
14639.09 14639.18
14639.09EA 82.4 EUR
14639.09 14639.18
Silver ≥99.9% (metals basis), flakes 80% <20 µm
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: Scott density 18-28g/in3, S.A. 1.0-2.2m2/g Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III
Merck Index: 12,08647