Silver ≥99.9% (metals basis), soluble 80% <20 µm
Supplier: Thermo Fisher Scientific
14755.09 14755.50
14755.09EA 125 EUR
14755.09 14755.50
Silver ≥99.9% (metals basis), soluble 80% <20 µm
Silver
Purity: 99.9% CAS No.: 7440-22-4 Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397
Suitable for manufacturing metallizing preparations and conductive silicone rubbers
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9 III
Merck Index: 12,08647