Silver ≥99.9% (metals basis), foil, annealed, Thickness: 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
41453.FI 41453.GH
41453.FIEA 270 EUR
41453.FI 41453.GH
Silver ≥99.9% (metals basis), foil, annealed, Thickness: 2.0 mm (0.08 in)
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈13g/25x25mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647