Silver ≥99.9% (metals basis), spherical powder -635 mesh
Supplier: Thermo Fisher Scientific
41600.09 41600.18 41600.30
41600.09EA 139 EUR
41600.09 41600.18 41600.30
Silver ≥99.9% (metals basis), spherical powder -635 mesh
Silver
Purity: 99.9% CAS No.: 7440-22-4 Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 961.9 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III