Silver ≥99.9% (metals basis), wool
Supplier: Thermo Fisher Scientific
42860.03 42860.06 42860.14
42860.03EA 37.1 EUR
42860.03 42860.06 42860.14
Silver ≥99.9% (metals basis), wool
Silver
Purity: 99.9+% CAS No.: 7440-22-4 Physical Form: Approximately 0.05mm dia fibers Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647