Silver, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
265608-41G 265608-8.2G
SIAL265608-41GEA 305 EUR
SIAL265608-41G SIAL265608-8.2G
Silver, Sigma-Aldrich®
Silver
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647