Cadmium ≥99.95% (metals basis), tear drop shot -5/+20 mesh
Supplier: Thermo Fisher Scientific
Danger
10122.18 10122.30 10122.A1
10122.18EA 53.7 EUR
10122.18 10122.30 10122.A1
Cadmium ≥99.95% (metals basis), tear drop shot -5/+20 mesh
Cadmium
Purity: 99.95% CAS No.: 7440-43-9 MDL No.: MFCD00010914
Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649