Cadmium ≥99.998% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Danger
11910.G1 11910.G5
11910.G1EA 72.1 EUR
11910.G1 11910.G5
Cadmium ≥99.998% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Cadmium
Purity: 99.998% CAS No.: 7440-43-9 Physical Form: ≈1.7g/m MDL No.: MFCD00010914
Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649