Mercury
Supplier: Roth Carl
Danger
7593.1 7593.2
ROTH7593.1EA 0 EUR
ROTH7593.1 ROTH7593.2
Mercury
Mercury
Formula: Hg
MW: 200.59 g/mol
Boiling Pt: 356.5
Melting Pt: –39 °C
Density: 13.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011035
CAS Number: 7439-97-6
EINECS: 231-106-7
UN: 2809
ADR: 8 III
Merck Index: 13,05925