Zinc, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
267635-100G 267635-25G
SIAL267635-100GEA 440 EUR
SIAL267635-100G SIAL267635-25G
Zinc, Sigma-Aldrich®
Zinc
Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255