Zinc, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
267929-1.4G 267929-5.6G
SIAL267929-1.4GEA 169 EUR
SIAL267929-1.4G SIAL267929-5.6G
Zinc, Sigma-Aldrich®
Zinc
Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255