Zinc, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

402583-1KG 402583-250G
SIAL402583-1KGEA 110 EUR
SIAL402583-1KG SIAL402583-250G
Zinc, Sigma-Aldrich®
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR