Titanium ≥99.7% (metals basis), wire, Length: 91 cm (36 in), Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Warning
10397.KJ 10397.KM
10397.KJEA 149 EUR
10397.KJ 10397.KM
Titanium ≥99.7% (metals basis), wire, Length: 91 cm (36 in), Ø 2.0 mm (0.08 in)
Titanium
Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈12.9g/pc Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610