Titanium ≥99.2% (metals basis), foil, annealed, Thickness: 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Warning
10398.GH 10398.GW
10398.GHEA 87.2 EUR
10398.GH 10398.GW
Titanium ≥99.2% (metals basis), foil, annealed, Thickness: 2.0 mm (0.08 in)
Titanium
Purity: 99.2% CAS No.: 7440-32-6 Physical Form: ≈90g/100x100mm Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610