Titanium ≥99.7% (metals basis), foil, annealed, Thickness: 0.89 mm (0.035 in)
Supplier: Thermo Fisher Scientific
Warning
10399.GH 10399.GW 10399.ZB
10399.GHEA 54.4 EUR
10399.GH 10399.GW 10399.ZB
Titanium ≥99.7% (metals basis), foil, annealed, Thickness: 0.89 mm (0.035 in)
Titanium
Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈40g/100x100mm Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610