Titanium, gauze woven from wire 20 mesh, Ø 0.076 mm (0.003 in)
Supplier: Thermo Fisher Scientific
Warning
40918.FL 40918.HB
40918.FLEA 133 EUR
40918.FL 40918.HB
Titanium, gauze woven from wire 20 mesh, Ø 0.076 mm (0.003 in)
Titanium
CAS No.: 7440-32-6 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610