Titanium, gauze woven from wire 40 mesh, Ø 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
Warning
40920.FL 40920.GT 40920.HB
40920.FLEA 139 EUR
40920.FL 40920.GT 40920.HB
Titanium, gauze woven from wire 40 mesh, Ø 0.127 mm (0.005 in)
Titanium
CAS No.: 7440-32-6 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610