Titanium ≥99.9% (metals basis), sponge ≤ 3 mm (0.12 in)
Supplier: Thermo Fisher Scientific
Warning
42459.30 42459.A1 42459.A7
42459.30EA 166 EUR
42459.30 42459.A1 42459.A7
Titanium ≥99.9% (metals basis), sponge ≤ 3 mm (0.12 in)
Titanium
Purity: 99.9% CAS No.: 7440-32-6 Odour: Odourless UN No.: 2878 MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2878
ADR: 4.1 III
Merck Index: 12,09610