Titanium ≥99.99% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Warning
43676.GH 43676.FF 43676.FI
43676.GHEA 1330 EUR
43676.GH 43676.FF 43676.FI
Titanium ≥99.99% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Titanium
Purity: 99.99+% CAS No.: 7440-32-6 Physical Form: ≈1.4g/25x25mm Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610