Titanium ≥99.99% (metals basis), powder -325 mesh, dehydrided
Supplier: Thermo Fisher Scientific
Danger
36679.14 36679.22
36679.14EA 245 EUR
36679.14 36679.22
Titanium ≥99.99% (metals basis), powder -325 mesh, dehydrided
Titanium
Purity: 99.99% CAS No.: 7440-32-6 Odour: Odourless UN No.: 2546 MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2 II
Merck Index: 12,09610