Zirconium ≥99.94% (metals basis excluding Hf), foil, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
00417.FF 00417.FQ
00417.FFEA 93.9 EUR
00417.FF 00417.FQ
Zirconium ≥99.94% (metals basis excluding Hf), foil, Thickness: 0.25 mm (0.01 in)
Zirconium
Purity: 99.94% CAS No.: 7440-67-7 Physical Form: ≈1g/25x25mm Odour: Odourless MDL No.: MFCD00011303
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9