Zirconium ≥99.5% (Zr, Hf), sponge 0.8-25.4 mm (0.03-1.0 in)
Supplier: Thermo Fisher Scientific
Danger
10441.18 10441.30 10441.A1
10441.18EA 74.2 EUR
10441.18 10441.30 10441.A1
Zirconium ≥99.5% (Zr, Hf), sponge 0.8-25.4 mm (0.03-1.0 in)
Zirconium
Purity: 99.5% CAS No.: 7440-67-7 Odour: Odourless UN No.: 2008 MDL No.: MFCD00011303
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9
UN: 2008
ADR: 4.2 III