Zirconium ≥99.2% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
10442.BQ 10442.BV
10442.BQEA 99.9 EUR
10442.BQ 10442.BV
Zirconium ≥99.2% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 12.7 mm (0.5 in)
Zirconium
Purity: 99.2+% CAS No.: 7440-67-7 Physical Form: ≈8.24g/cm, Temper: soft Odour: Odourless MDL No.: MFCD00011303
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9