Zirconium ≥99.2% (metals basis excluding Hf), foil (max. 4.5% Hf), annealed, Thickness: 1.0 mm (0.039 in)
Supplier: Thermo Fisher Scientific
10590.FI 10590.GH 10590.HP
10590.FIEA 72.1 EUR
10590.FI 10590.GH 10590.HP
Zirconium ≥99.2% (metals basis excluding Hf), foil (max. 4.5% Hf), annealed, Thickness: 1.0 mm (0.039 in)
Zirconium
Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈16.3g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00011303
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9