Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
42509.14 42509.22
42509.14EA 55.8 EUR
42509.14 42509.22
Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Zirconium
Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈1.31g/slug Odour: Odourless MDL No.: MFCD00011303
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9