Zirconium 99,5 Metals basis exHf, slug Hf_about3, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
42510.09 42510.18
42510.09EA 45.2 EUR
42510.09 42510.18
Zirconium 99,5 Metals basis exHf, slug Hf_about3, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Zirconium
Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈0.33g/slug Odour: Odourless MDL No.: MFCD00011303
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9