Zirconium ≥99.5% (metals basis excluding Hf), foil, annealed, Thickness: 0.711 mm (0.028 in)
Supplier: Thermo Fisher Scientific
44253.GH 44253.HP
44253.GHEA 199 EUR
44253.GH 44253.HP
Zirconium ≥99.5% (metals basis excluding Hf), foil, annealed, Thickness: 0.711 mm (0.028 in)
Zirconium
Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈46.2g/100x100mm, Temper: soft Odour: Odourless MDL No.: MFCD00011303
Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9