Zirconium ≥99.8% (metals basis), foil, annealed, Thickness: 0.20 mm (0.008 in)
Supplier: Thermo Fisher Scientific
44752.GI 44752.GR 44752.IB
44752.GIEA 96.1 EUR
44752.GI 44752.GR 44752.IB
Zirconium ≥99.8% (metals basis), foil, annealed, Thickness: 0.20 mm (0.008 in)
Zirconium
Purity: 99.8% CAS No.: 7440-67-7 Physical Form: ≈20.3g/100x125mm, Temper: soft Odour: Odourless MDL No.: MFCD00011303
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9