Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
10261.CR 10261.CS 10261.CT
10261.CREA 64.3 EUR
10261.CR 10261.CS 10261.CT
Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Niobium
Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈5.44g/5x10cm, Temper: soft Odour: Odourless MDL No.: MFCD00011126
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5